Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HRASLS2
Entrez Gene ID
54979
Gene Name
HRAS like suppressor 2
Gene Aliases
PLA1/2-2
Chromosome Location
Chr.11: 63552759 - 63565072 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017878.1 NP_060348.1 1 - 2 72 74
GenBank mRNA AK000563.1 - 1 - 2 72 74

View Full Product Details