Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLAMF8
Entrez Gene ID
56833
Gene Name
SLAM family member 8
Gene Aliases
BLAME, CD353, SBBI42
Chromosome Location
Chr.1: 159826650 - 159837497 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020125.2 NP_064510.1 4 - 5 993 89
XM_011509780.2 XP_011508082.2 3 - 4 600 89
XM_017001846.1 XP_016857335.1 4 - 5 880 89
GenBank mRNA AF144235.1 - 1 - 2 74 89
AF146761.1 - 4 - 5 871 89
AK074669.1 - 4 - 5 932 89
BC109194.2 - 4 - 5 809 89

View Full Product Details