The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020903.2 | NP_065954.1 | 4 - 4 | 1146 | 62 |
GenBank mRNA | AF124433.1 | - | 4 - 4 | 1158 | 62 |
AF229438.1 | - | 1 - 1 | 823 | 62 | |
AK308285.1 | - | 4 - 4 | 1244 | 62 | |
BC130394.1 | - | 2 - 2 | 906 | 62 | |
BC130398.1 | - | 2 - 2 | 906 | 62 |