Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MBNL1
Entrez Gene ID
4154
Gene Name
muscleblind like splicing regulator 1
Gene Aliases
EXP, MBNL
Chromosome Location
Chr.3: 152243656 - 152465780 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001314057.1 NP_001300986.1 4 - 5 981 55
NM_021038.4 NP_066368.2 3 - 4 1766 55
NM_207292.2 NP_997175.1 3 - 4 1766 55
NM_207293.1 NP_997176.1 2 - 3 1141 55
NM_207296.1 NP_997179.1 2 - 3 1141 55
NM_207297.1 NP_997180.1 2 - 3 1141 55
XM_005247457.4 XP_005247514.1 4 - 5 1334 55
XM_005247458.4 XP_005247515.1 4 - 5 2588 55
XM_005247460.4 XP_005247517.1 4 - 5 1297 55
XM_005247461.4 XP_005247518.1 4 - 5 1334 55
XM_005247462.4 XP_005247519.1 4 - 5 1334 55
XM_005247463.4 XP_005247520.1 4 - 5 1334 55
XM_005247464.4 XP_005247521.1 3 - 4 1392 55
XM_005247465.4 XP_005247522.1 4 - 5 1334 55
XM_005247466.4 XP_005247523.1 3 - 4 1369 55
XM_005247467.4 XP_005247524.1 4 - 5 1334 55
XM_005247468.4 XP_005247525.1 4 - 5 1334 55
XM_005247471.4 XP_005247528.1 3 - 4 1364 55
XM_005247474.3 XP_005247531.1 2 - 3 252 55
XM_005247475.3 XP_005247532.1 3 - 4 575 55
XM_005247476.4 XP_005247533.1 2 - 3 342 55
XM_005247477.4 XP_005247534.1 2 - 3 341 55
XM_006713639.1 XP_006713702.1 4 - 5 1316 55
XM_011512848.2 XP_011511150.1 4 - 5 938 55
XM_017006423.1 XP_016861912.1 3 - 4 1585 55
XM_017006424.1 XP_016861913.1 3 - 4 1412 55
XM_017006425.1 XP_016861914.1 3 - 4 1414 55
XM_017006426.1 XP_016861915.1 3 - 4 1412 55
XM_017006427.1 XP_016861916.1 3 - 4 1559 55
XM_017006428.1 XP_016861917.1 3 - 4 1223 55
XM_017006429.1 XP_016861918.1 3 - 4 1306 55
XM_017006430.1 XP_016861919.1 3 - 4 1370 55
XM_017006431.1 XP_016861920.1 3 - 4 1447 55
XM_017006432.1 XP_016861921.1 3 - 4 1369 55
XM_017006433.1 XP_016861922.1 3 - 4 1369 55
XM_017006434.1 XP_016861923.1 3 - 4 1375 55
XM_017006435.1 XP_016861924.1 3 - 4 382 55
XM_017006436.1 XP_016861925.1 3 - 4 2495 55
XM_017006437.1 XP_016861926.1 3 - 4 1550 55
XM_017006439.1 XP_016861928.1 3 - 4 575 55
XM_017006440.1 XP_016861929.1 2 - 3 309 55
XM_017006441.1 XP_016861930.1 2 - 3 342 55
XM_017006442.1 XP_016861931.1 2 - 3 445 55
XM_017006443.1 XP_016861932.1 2 - 3 453 55
GenBank mRNA AB007888.1 - 3 - 4 1766 55
AF401998.1 - 2 - 3 352 55
AF497718.1 - 2 - 3 352 55
AF497719.1 - 2 - 3 352 55
AJ308400.1 - 2 - 3 381 55
AK304089.1 - 1 - 2 61 55
AK307420.1 - 1 - 2 61 55
BC043493.1 - 3 - 4 1271 55
BC050535.1 - 4 - 5 502 55
Y13829.1 - 2 - 3 863 55

View Full Product Details