The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001129758.1 | NP_001123230.1 | 4 - 5 | 855 | 63 |
NM_022357.3 | NP_071752.3 | 4 - 5 | 855 | 63 | |
GenBank mRNA | AJ291679.1 | - | 4 - 5 | 769 | 63 |
AK057401.1 | - | 4 - 5 | 855 | 63 | |
AY358390.1 | - | 4 - 5 | 778 | 63 | |
BC037243.1 | - | 4 - 5 | 767 | 63 | |
BC057789.1 | - | 4 - 5 | 796 | 63 |