Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MMP16
- Entrez Gene ID
-
4325
- Gene Name
- matrix metallopeptidase 16
- Gene Aliases
-
C8orf57, MMP-X2, MT-MMP2, MT-MMP3, MT3-MMP
- Chromosome Location
-
Chr.8: 88037232 - 88327488 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
D83647.1 |
- |
7 - 8 |
1222 |
90 |
View Full Product Details