Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GAR1
Entrez Gene ID
54433
Gene Name
GAR1 ribonucleoprotein
Gene Aliases
NOLA1
Chromosome Location
Chr.4: 109815510 - 109824737 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018983.3 NP_061856.1 1 - 2 295 100
GenBank mRNA AA311919.1 - 1 - 2 261 100
AJ276003.1 - 1 - 2 100
CN428426.1 - 1 - 2 260 100

View Full Product Details