Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PER2
Entrez Gene ID
8864
Gene Name
period circadian clock 2
Gene Aliases
FASPS, FASPS1
Chromosome Location
Chr.2: 238244038 - 238290102 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022817.2 NP_073728.1 9 - 10 1281 135
XM_005246111.4 XP_005246168.1 9 - 10 1267 135
XM_006712824.3 XP_006712887.1 9 - 10 1219 135
GenBank mRNA AB002345.2 - 9 - 10 1166 135
AK294876.1 - 9 - 10 1281 135
AK304108.1 - 9 - 10 1267 135

View Full Product Details