Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 185149969-185150428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A4
Entrez Gene ID
291
Gene Name
solute carrier family 25 member 4
Gene Aliases
AAC1, ANT, ANT 1, ANT1, MTDPS12, PEO2, PEO3, PEOA2, T1
UniGene
Hs.246506
Gene Chromosome Location
Chr.4: 185143263-185150384 on Build GRCh38
Transcripts
NM_001151.3

SNP Detail

SNP ID Chromosome Location
rs1037150884 Chr.4: 185150174-185150174 on Build GRCh38
rs962471297 Chr.4: 185150200-185150200 on Build GRCh38
rs559209892 Chr.4: 185150175-185150175 on Build GRCh38
rs752130203 Chr.4: 185150288-185150288 on Build GRCh38
rs973541096 Chr.4: 185150203-185150203 on Build GRCh38
rs946805697 Chr.4: 185150281-185150281 on Build GRCh38
rs938018765 Chr.4: 185150106-185150106 on Build GRCh38
rs1052549725 Chr.4: 185150067-185150067 on Build GRCh38
rs773137655 Chr.4: 185149996-185149996 on Build GRCh38
rs911858806 Chr.4: 185150263-185150263 on Build GRCh38
rs979478625 Chr.4: 185150294-185150294 on Build GRCh38
rs182549219 Chr.4: 185150166-185150166 on Build GRCh38
rs932224312 Chr.4: 185150234-185150234 on Build GRCh38
rs897353935 Chr.4: 185150179-185150179 on Build GRCh38
rs147626367 Chr.4: 185150085-185150085 on Build GRCh38
rs528416282 Chr.4: 185150217-185150217 on Build GRCh38
rs925245760 Chr.4: 185150100-185150100 on Build GRCh38
rs1003876425 Chr.4: 185150395-185150395 on Build GRCh38
rs116996106 Chr.4: 185150081-185150081 on Build GRCh38
rs752912280 Chr.4: 185150150-185150150 on Build GRCh38
rs753481356 Chr.4: 185150381-185150381 on Build GRCh38
rs755875204 Chr.4: 185150089-185150089 on Build GRCh38
rs533345524 Chr.4: 185150178-185150178 on Build GRCh38
rs766108137 Chr.4: 185150092-185150092 on Build GRCh38
rs932276798 Chr.4: 185149995-185149995 on Build GRCh38
rs925362228 Chr.4: 185150307-185150307 on Build GRCh38
rs1055529077 Chr.4: 185150347-185150347 on Build GRCh38
rs558959854 Chr.4: 185150042-185150042 on Build GRCh38
rs1043856025 Chr.4: 185150099-185150099 on Build GRCh38
rs187103619 Chr.4: 185150247-185150247 on Build GRCh38
rs530438950 Chr.4: 185150279-185150279 on Build GRCh38
rs905038209 Chr.4: 185150332-185150332 on Build GRCh38
rs1003844327 Chr.4: 185150362-185150362 on Build GRCh38
rs1016595384 Chr.4: 185150398-185150398 on Build GRCh38
rs890491299 Chr.4: 185150017-185150017 on Build GRCh38
rs922129353 Chr.4: 185150229-185150229 on Build GRCh38
rs191872006 Chr.4: 185150386-185150386 on Build GRCh38
rs765546292 Chr.4: 185150143-185150143 on Build GRCh38
rs549902846 Chr.4: 185150376-185150376 on Build GRCh38
rs1052898444 Chr.4: 185150274-185150274 on Build GRCh38
rs140827011 Chr.4: 185150093-185150093 on Build GRCh38
rs1055343451 Chr.4: 185150119-185150119 on Build GRCh38
rs376762770 Chr.4: 185150090-185150090 on Build GRCh38
rs938149344 Chr.4: 185150333-185150333 on Build GRCh38
rs759840559 Chr.4: 185150137-185150137 on Build GRCh38
rs189929266 Chr.4: 185150086-185150086 on Build GRCh38
rs754761545 Chr.4: 185150387-185150387 on Build GRCh38
rs539027516 Chr.4: 185150041-185150041 on Build GRCh38
rs1017307771 Chr.4: 185150253-185150253 on Build GRCh38
rs966551646 Chr.4: 185150291-185150291 on Build GRCh38
rs886059277 Chr.4: 185150248-185150248 on Build GRCh38
rs764517871 Chr.4: 185150264-185150264 on Build GRCh38
rs564927574 Chr.4: 185150277-185150277 on Build GRCh38
rs896728617 Chr.4: 185150139-185150139 on Build GRCh38
rs542420297 Chr.4: 185150128-185150128 on Build GRCh38
rs1035069248 Chr.4: 185150114-185150114 on Build GRCh38

View Full Product Details