Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC25A28
Entrez Gene ID
81894
Gene Name
solute carrier family 25 member 28
Gene Aliases
MFRN2, MRS3/4, MRS4L, NPD016
Chromosome Location
Chr.10: 99610518 - 99659329 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031212.3 NP_112489.3 4 - 4 958 69
XM_005270208.1 XP_005270265.1 4 - 4 948 69
XM_005270211.4 XP_005270268.1 4 - 4 562 69
XM_006718005.3 XP_006718068.1 4 - 4 622 69
XM_011540239.2 XP_011538541.1 4 - 4 844 69
XM_011540240.1 XP_011538542.1 5 - 5 2094 69
XM_017016741.1 XP_016872230.1 4 - 4 2030 69
XM_017016742.1 XP_016872231.1 4 - 4 1851 69
GenBank mRNA AF267854.1 - 1 - 1 645 69
AF327402.1 - 4 - 4 858 69
AF327403.1 - 4 - 4 1299 69
AJ303077.1 - 4 - 4 858 69
AJ303078.1 - 4 - 4 1299 69
AK056782.1 - 1 - 1 998 69
AL831943.1 - 1 - 1 520 69
BC047312.1 - 4 - 4 905 69
BC058937.1 - 4 - 4 867 69
BC076399.1 - 4 - 4 898 69
BC094821.1 - 4 - 4 641 69

View Full Product Details