Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- AGTR1
- Entrez Gene ID
-
185
- Gene Name
- angiotensin II receptor type 1
- Gene Aliases
-
AG2S, AGTR1B, AT1, AT1AR, AT1B, AT1BR, AT1R, AT2R1, HAT1R
- Chromosome Location
-
Chr.3: 148697871 - 148743003 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_031850.3 |
NP_114038.4 |
2 - 3 |
340 |
67 |
View Full Product Details