The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_103511.1 | 3 - 4 | 712 | 83 | |
XM_006713783.3 | XP_006713846.1 | 2 - 3 | 2116 | 83 | |
XM_017007311.1 | XP_016862800.1 | 4 - 5 | 2295 | 83 | |
XM_017007312.1 | XP_016862801.1 | 3 - 4 | 2350 | 83 | |
XM_017007313.1 | XP_016862802.1 | 3 - 4 | 2351 | 83 | |
GenBank mRNA | AK292958.1 | - | 2 - 3 | 376 | 83 |
AK310829.1 | - | 2 - 3 | 367 | 83 | |
AL136815.1 | - | 1 - 2 | 143 | 83 | |
BC024239.2 | - | 3 - 4 | 482 | 83 | |
BG722744.1 | - | 3 - 4 | 487 | 83 | |
BM705017.1 | - | 3 - 4 | 333 | 83 |