The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135021.1 | NP_001128493.1 | 11 - 12 | 1270 | 98 |
NM_001135022.1 | NP_001128494.1 | 10 - 11 | 1086 | 98 | |
NM_001135023.1 | NP_001128495.1 | 9 - 10 | 1042 | 98 | |
NM_032213.4 | NP_115589.2 | 8 - 9 | 947 | 98 | |
XM_005264596.3 | XP_005264653.1 | 10 - 11 | 1086 | 98 | |
XM_005264597.3 | XP_005264654.1 | 9 - 10 | 1368 | 98 | |
XM_005264598.3 | XP_005264655.1 | 9 - 10 | 991 | 98 | |
XM_017005078.1 | XP_016860567.1 | 9 - 10 | 942 | 98 | |
XM_017005079.1 | XP_016860568.1 | 8 - 9 | 898 | 98 | |
GenBank mRNA | BC001942.1 | - | 8 - 9 | 864 | 98 |
BC010991.2 | - | 10 - 11 | 1018 | 98 | |
BC018666.2 | - | 8 - 9 | 864 | 98 | |
BC054504.1 | - | 8 - 9 | 2814 | 98 | |
BC112324.1 | - | 9 - 10 | 1005 | 98 | |
DQ256726.1 | - | 6 - 7 | 843 | 98 | |
DQ256727.1 | - | 7 - 8 | 613 | 98 |