The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011537.2 | NP_001011537.2 | 8 - 9 | 889 | 127 |
NM_032288.6 | NP_115664.2 | 7 - 8 | 948 | 127 | |
NR_027840.1 | 7 - 8 | 1059 | 127 | ||
GenBank mRNA | AJ344094.1 | - | 7 - 8 | 794 | 127 |
AK296283.1 | - | 6 - 7 | 650 | 127 | |
AK302546.1 | - | 7 - 8 | 1059 | 127 | |
AK303462.1 | - | 8 - 9 | 889 | 127 | |
AK315019.1 | - | 7 - 8 | 797 | 127 | |
AK316308.1 | - | 7 - 8 | 689 | 127 | |
AL136558.1 | - | 7 - 8 | 801 | 127 | |
BC035006.2 | - | 7 - 8 | 951 | 127 | |
BC039734.1 | - | 7 - 8 | 858 | 127 | |
CR533502.1 | - | 7 - 8 | 729 | 127 |