The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032359.3 | NP_115735.2 | 1 - 2 | 214 | 80 |
GenBank mRNA | AK124387.1 | - | 1 - 2 | 181 | 80 |
AK291392.1 | - | 1 - 2 | 186 | 80 | |
AK312483.1 | - | 1 - 2 | 214 | 80 | |
BC006475.1 | - | 1 - 2 | 211 | 80 | |
BC006512.2 | - | 1 - 2 | 211 | 80 |