The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145459.1 | NP_001138931.1 | 2 - 3 | 377 | 57 |
NM_032495.5 | NP_115884.4 | 3 - 4 | 731 | 57 | |
NM_139211.4 | NP_631957.1 | 2 - 3 | 333 | 57 | |
NM_139212.3 | NP_631958.1 | 2 - 3 | 606 | 57 | |
XM_017008728.1 | XP_016864217.1 | 2 - 3 | 368 | 57 | |
XM_017008729.1 | XP_016864218.1 | 2 - 3 | 250 | 57 | |
XM_017008730.1 | XP_016864219.1 | 2 - 3 | 298 | 57 | |
XM_017008731.1 | XP_016864220.1 | 2 - 3 | 333 | 57 | |
XM_017008732.1 | XP_016864221.1 | 3 - 4 | 609 | 57 | |
XM_017008733.1 | XP_016864222.1 | 2 - 3 | 173 | 57 | |
XM_017008734.1 | XP_016864223.1 | 2 - 3 | 229 | 57 | |
GenBank mRNA | AB059408.1 | - | 2 - 3 | 291 | 57 |
AB059409.1 | - | 2 - 3 | 174 | 57 | |
AB059410.2 | - | 2 - 3 | 183 | 57 | |
AF454763.1 | - | 2 - 3 | 196 | 57 | |
AF492675.1 | - | 2 - 3 | 580 | 57 | |
AF492676.1 | - | 3 - 4 | 312 | 57 | |
AF492677.1 | - | 3 - 4 | 356 | 57 | |
AF492680.1 | - | 2 - 3 | 332 | 57 | |
AF492681.1 | - | 2 - 3 | 295 | 57 | |
AK289707.1 | - | 2 - 3 | 275 | 57 | |
AW955211.1 | - | 2 - 3 | 299 | 57 | |
BC014225.2 | - | 1 - 2 | 446 | 57 | |
CB996541.1 | - | 2 - 3 | 369 | 57 | |
EB386856.1 | - | 2 - 3 | 327 | 57 |