The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080156.2 | NP_001073625.1 | 15 - 16 | 1904 | 78 |
NM_001319850.1 | NP_001306779.1 | 20 - 21 | 2487 | 78 | |
NM_001319851.1 | NP_001306780.1 | 14 - 15 | 2180 | 78 | |
NM_001319852.1 | NP_001306781.1 | 15 - 16 | 1961 | 78 | |
NM_032496.3 | NP_115885.2 | 17 - 18 | 2185 | 78 | |
XM_005269083.2 | XP_005269140.1 | 15 - 16 | 1626 | 78 | |
XM_005269084.2 | XP_005269141.1 | 14 - 15 | 1559 | 78 | |
XM_005269085.2 | XP_005269142.1 | 14 - 15 | 1533 | 78 | |
XM_011538656.2 | XP_011536958.1 | 19 - 20 | 2222 | 78 | |
XM_011538657.2 | XP_011536959.1 | 16 - 17 | 2063 | 78 | |
XM_011538658.2 | XP_011536960.1 | 16 - 17 | 2005 | 78 | |
XM_017019800.1 | XP_016875289.1 | 16 - 17 | 2499 | 78 | |
XM_017019801.1 | XP_016875290.1 | 15 - 16 | 1569 | 78 | |
XM_017019802.1 | XP_016875291.1 | 14 - 15 | 1502 | 78 | |
GenBank mRNA | AB030239.1 | - | 15 - 16 | 1656 | 78 |
AB051853.1 | - | 17 - 18 | 2163 | 78 | |
AK057573.1 | - | 14 - 15 | 1833 | 78 | |
AK131415.1 | - | 20 - 21 | 2393 | 78 | |
AK301095.1 | - | 15 - 16 | 1556 | 78 | |
AL713803.1 | - | 10 - 11 | 1248 | 78 | |
BC006107.1 | - | 20 - 21 | 2487 | 78 | |
BC011820.1 | - | 15 - 16 | 1630 | 78 |