The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318918.1 | NP_001305847.1 | 4 - 5 | 1173 | 141 |
NM_032575.2 | NP_115964.2 | 2 - 3 | 409 | 141 | |
XM_005255641.4 | XP_005255698.1 | 3 - 4 | 474 | 141 | |
GenBank mRNA | AF325914.1 | - | 2 - 3 | 409 | 141 |
AK126918.1 | - | 4 - 5 | 1173 | 141 |