The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032833.4 | NP_116222.4 | 1 - 2 | 2329 | 92 |
GenBank mRNA | AK027650.1 | - | 1 - 2 | 2329 | 92 |
AK222877.1 | - | 1 - 2 | 2330 | 92 | |
AK303535.1 | - | 2 - 3 | 1883 | 92 | |
AL833746.1 | - | 1 - 2 | 1006 | 92 | |
BC065280.1 | - | 1 - 2 | 2303 | 92 |