The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032856.3 | NP_116245.2 | 7 - 8 | 951 | 62 |
NR_130944.1 | 6 - 7 | 1388 | 62 | ||
NR_130945.1 | 8 - 9 | 1064 | 62 | ||
NR_130946.1 | 7 - 8 | 958 | 62 | ||
NR_130947.1 | 6 - 7 | 862 | 62 | ||
GenBank mRNA | AF161382.1 | - | 7 - 8 | 882 | 62 |
AK027200.1 | - | 2 - 3 | 1565 | 62 | |
AK027794.1 | - | 7 - 8 | 888 | 62 | |
AK055578.1 | - | 6 - 7 | 1388 | 62 | |
AK055729.1 | - | 6 - 7 | 1470 | 62 | |
AK225068.1 | - | 7 - 8 | 891 | 62 | |
AK225199.1 | - | 7 - 8 | 888 | 62 | |
BC014115.2 | - | 7 - 8 | 889 | 62 | |
BC020252.2 | - | 6 - 7 | 1051 | 62 | |
BC050648.1 | - | 7 - 8 | 908 | 62 | |
BC063392.1 | - | 7 - 8 | 919 | 62 |