The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032961.2 | NP_116586.1 | 4 - 5 | 3957 | 86 |
GenBank mRNA | AB037821.1 | - | 4 - 5 | 3932 | 86 |
AK024636.1 | - | 4 - 5 | 1221 | 86 | |
AL157470.1 | - | 4 - 5 | 1014 | 86 | |
BC111560.1 | - | 4 - 5 | 3295 | 86 |