Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CCL15
- Entrez Gene ID
-
6359
- Gene Name
- C-C motif chemokine ligand 15
- Gene Aliases
-
HCC-2, HMRP-2B, LKN-1, LKN1, MIP-1 delta, MIP-1D, MIP-5, MRP-2B, NCC-3, NCC3, SCYA15, SCYL3, SY15
- Chromosome Location
-
Chr.17: 35997582 - 36002038 on Build GRCh38
- Gene Symbol
- CCL15-CCL14
- Entrez Gene ID
-
348249
- Gene Name
- CCL15-CCL14 readthrough (NMD candidate)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 35983656 - 36002038 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027921.3 |
|
2 - 3 |
677 |
111 |
NR_027922.3 |
|
2 - 3 |
677 |
111 |
GenBank mRNA |
BC140941.1 |
- |
2 - 3 |
260 |
111 |
View Full Product Details