The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145811.1 | NP_001139283.1 | 5 - 6 | 903 | 81 |
NM_001145819.1 | NP_001139291.1 | 5 - 6 | 784 | 81 | |
NM_017508.2 | NP_059978.1 | 5 - 6 | 925 | 81 | |
NM_033326.3 | NP_201583.2 | 5 - 6 | 790 | 81 | |
GenBank mRNA | AF309034.1 | - | 5 - 6 | 773 | 81 |
AL136780.1 | - | 5 - 6 | 904 | 81 | |
BC047064.1 | - | 5 - 6 | 780 | 81 |