Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADRA2A
Entrez Gene ID
150
Gene Name
adrenoceptor alpha 2A
Gene Aliases
ADRA2, ADRA2R, ADRAR, ALPHA2AAR, ZNF32
Chromosome Location
Chr.10: 111077032 - 111080907 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000681.3 NP_000672.3 1 - 1 1960 116
GenBank mRNA AF284095.1 - 1 - 1 1829 116
BC035047.1 - 1 - 1 1163 116
BC050414.1 - 1 - 1 1345 116

View Full Product Details