Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IMPDH1
Entrez Gene ID
3614
Gene Name
inosine monophosphate dehydrogenase 1
Gene Aliases
IMPD, IMPD1, IMPDH-I, LCA11, RP10, sWSS2608
Chromosome Location
Chr.7: 128392277 - 128409988 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000883.3 NP_000874.2 1 - 2 232 62
NM_001142576.1 NP_001136048.1 1 - 2 232 62
XM_017012171.1 XP_016867660.1 1 - 2 232 62
XM_017012172.1 XP_016867661.1 1 - 2 232 62
GenBank mRNA DC326380.1 - 1 - 2 213 62
J05272.1 - 1 - 2 495 62

View Full Product Details