Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC3
Entrez Gene ID
7267
Gene Name
tetratricopeptide repeat domain 3
Gene Aliases
DCRR1, RNF105, TPRDIII
Chromosome Location
Chr.21: 37073184 - 37203118 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001894.1 NP_001001894.1 2 - 3 253 96
NM_001320703.1 NP_001307632.1 2 - 3 459 96
NM_001320704.1 NP_001307633.1 2 - 3 449 96
NM_003316.3 NP_003307.3 2 - 3 1544 96
XM_005261048.1 XP_005261105.1 2 - 3 311 96
XM_005261049.1 XP_005261106.1 2 - 3 234 96
XM_005261050.4 XP_005261107.1 2 - 3 3196 96
XM_005261052.1 XP_005261109.1 2 - 3 234 96
XM_005261054.3 XP_005261111.1 2 - 3 3188 96
XM_011529740.2 XP_011528042.1 1 - 2 159 96
XM_011529742.2 XP_011528044.2 2 - 3 256 96
XM_017028458.1 XP_016883947.1 2 - 3 250 96
XM_017028459.1 XP_016883948.1 2 - 3 249 96
XM_017028460.1 XP_016883949.1 2 - 3 248 96
XM_017028461.1 XP_016883950.1 2 - 3 311 96
XM_017028462.1 XP_016883951.1 2 - 3 234 96
XM_017028463.1 XP_016883952.1 2 - 3 311 96
GenBank mRNA AK125473.1 - 2 - 3 2896 96
AK291992.1 - 2 - 3 361 96
BC063033.1 - 2 - 3 271 96
BC092466.1 - 2 - 3 256 96
BC137345.1 - 2 - 3 683 96
D83077.1 - 2 - 3 1544 96
D84294.1 - 1 - 2 1617 96
D84295.1 - 1 - 2 1617 96

View Full Product Details