Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RRBP1
Entrez Gene ID
6238
Gene Name
ribosome binding protein 1
Gene Aliases
ES/130, ES130, RRp, hES
Chromosome Location
Chr.20: 17613678 - 17682283 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042576.1 NP_001036041.1 9 - 10 1665 67
NM_004587.2 NP_004578.2 8 - 9 1588 67
GenBank mRNA AB037819.2 - - 3344 67
AB287347.2 - 5 - 6 3000 67
AF006751.1 - 8 - 9 1381 67
AK309197.1 - 3 - 4 327 67
AL833822.1 - 6 - 7 1376 67
BC000099.2 - 3 - 4 346 67
BC020482.1 - 5 - 6 613 67
BC080526.1 - 4 - 5 472 67
BC128577.1 - 8 - 9 1774 67
BC128578.1 - 7 - 8 808 67

View Full Product Details