The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004975.3 | NP_004966.1 | 1 - 2 | 728 | 68 |
XM_006723784.3 | XP_006723847.1 | 2 - 3 | 822 | 68 | |
XM_011528799.2 | XP_011527101.1 | 2 - 3 | 851 | 68 | |
GenBank mRNA | AF026005.1 | - | 1 - 2 | 564 | 68 |
BC110572.1 | - | 2 - 3 | 429 | 68 | |
BC110573.1 | - | 2 - 3 | 429 | 68 | |
L02840.1 | - | 1 - 2 | 750 | 68 |