The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286099.1 | NP_001273028.1 | 3 - 3 | 612 | 102 |
NM_005284.4 | NP_005275.1 | 2 - 2 | 503 | 102 | |
GenBank mRNA | AK295250.1 | - | 3 - 3 | 557 | 102 |
BC043414.2 | - | 1 - 1 | 291 | 102 | |
BC117298.1 | - | 1 - 1 | 389 | 102 | |
BC126301.1 | - | 1 - 1 | 391 | 102 | |
CF147782.1 | - | 1 - 1 | 341 | 102 | |
DA205492.1 | - | 2 - 2 | 503 | 102 | |
DA260869.1 | - | 2 - 2 | 448 | 102 | |
HQ709190.1 | - | 1 - 1 | 273 | 102 | |
HQ709191.1 | - | 1 - 1 | 273 | 102 |