The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006098.4 | NP_006089.1 | 1 - 2 | 216 | 66 |
GenBank mRNA | AK095666.1 | - | 1 - 2 | 214 | 66 |
AK222488.1 | - | 1 - 2 | 216 | 66 | |
AK301029.1 | - | 1 - 2 | 52 | 66 | |
AK301468.1 | - | 1 - 2 | 212 | 66 | |
AK303316.1 | - | 1 - 2 | 224 | 66 | |
BC000214.1 | - | 1 - 2 | 234 | 66 | |
BC000366.2 | - | 1 - 2 | 204 | 66 | |
BC010119.2 | - | 1 - 2 | 191 | 66 | |
BC014256.1 | - | 1 - 2 | 193 | 66 | |
BC014788.1 | - | 1 - 2 | 200 | 66 | |
BC017287.2 | - | 1 - 2 | 181 | 66 | |
BC019093.2 | - | 1 - 2 | 194 | 66 | |
BC019362.1 | - | 1 - 2 | 198 | 66 | |
BC021993.1 | - | 1 - 2 | 216 | 66 | |
BC029996.1 | - | 1 - 2 | 43 | 66 | |
BC032006.1 | - | 1 - 2 | 195 | 66 | |
BC035460.1 | - | 1 - 2 | 197 | 66 | |
CR456978.1 | - | 1 - 2 | 110 | 66 | |
CR541909.1 | - | 1 - 2 | 110 | 66 | |
GU727632.1 | - | 1 - 2 | 110 | 66 | |
JN104586.1 | - | 1 - 2 | 394 | 66 | |
M24194.1 | - | 1 - 2 | 205 | 66 |