The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193529.2 | NP_001180458.1 | 9 - 10 | 1217 | 69 |
NM_001193530.1 | NP_001180459.1 | 9 - 10 | 1193 | 69 | |
NM_006584.3 | NP_006575.2 | 10 - 11 | 1328 | 69 | |
XM_011524203.2 | XP_011522505.1 | 8 - 9 | 947 | 69 | |
XM_017024023.1 | XP_016879512.1 | 10 - 11 | 1231 | 69 | |
XM_017024024.1 | XP_016879513.1 | 7 - 8 | 1088 | 69 | |
GenBank mRNA | AK225728.1 | - | 10 - 11 | 1249 | 69 |
AK225729.1 | - | 9 - 10 | 1110 | 69 | |
AK301773.1 | - | 9 - 10 | 1106 | 69 | |
AK302344.1 | - | 9 - 10 | 1140 | 69 | |
BC026125.2 | - | 10 - 11 | 1221 | 69 | |
BC027591.1 | - | 10 - 11 | 1308 | 69 | |
CR933688.1 | - | 4 - 5 | 2857 | 69 | |
D78333.1 | - | 10 - 11 | 1253 | 69 |