The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006691.3 | NP_006682.2 | 1 - 2 | 373 | 68 |
GenBank mRNA | AF118108.1 | - | 1 - 2 | 177 | 68 |
AF127670.2 | - | 1 - 2 | 207 | 68 | |
AK075443.1 | - | 1 - 2 | 287 | 68 | |
AK312462.1 | - | 1 - 2 | 371 | 68 | |
AY358925.1 | - | 1 - 2 | 246 | 68 | |
BC026231.1 | - | 1 - 2 | 132 | 68 | |
DA924658.1 | - | 1 - 2 | 373 | 68 |