The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013432.4 | NP_038460.4 | 13 - 14 | 1669 | 85 |
XM_011517048.2 | XP_011515350.1 | 6 - 7 | 768 | 85 | |
XM_011517049.2 | XP_011515351.1 | 6 - 7 | 729 | 85 | |
XM_011517050.2 | XP_011515352.1 | 13 - 14 | 1728 | 85 | |
GenBank mRNA | AB209126.1 | - | 5 - 6 | 2629 | 85 |
BC008782.2 | - | 12 - 13 | 1641 | 85 |