Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PELO
Entrez Gene ID
53918
Gene Name
pelota homolog (Drosophila)
Gene Aliases
CGI-17, PRO1770
Chromosome Location
Chr.5: 52787940 - 52802617 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF118075.2 - 1 - 1 364 86

View Full Product Details