Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BFAR
Entrez Gene ID
51283
Gene Name
bifunctional apoptosis regulator
Gene Aliases
BAR, RNF47
Chromosome Location
Chr.16: 14632811 - 14669236 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016561.2 NP_057645.1 2 - 3 545 103
GenBank mRNA AF173003.1 - 2 - 3 417 103
AK291114.1 - 2 - 3 353 103
AK297067.1 - 2 - 3 353 103
AK313890.1 - 1 - 2 353 103
BC003054.1 - 2 - 3 376 103

View Full Product Details