The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032167.4 | NP_115543.3 | 2 - 3 | 144 | 61 |
XM_005255682.3 | XP_005255739.1 | 2 - 3 | 169 | 61 | |
XM_005255683.3 | XP_005255740.1 | 2 - 3 | 185 | 61 | |
XM_011522738.2 | XP_011521040.1 | 2 - 3 | 316 | 61 | |
XM_011522741.2 | XP_011521043.1 | 2 - 3 | 300 | 61 | |
XM_011522743.2 | XP_011521045.1 | 2 - 3 | 304 | 61 | |
XM_011522744.2 | XP_011521046.1 | 2 - 3 | 175 | 61 | |
XM_017023871.1 | XP_016879360.1 | 2 - 3 | 313 | 61 | |
XM_017023872.1 | XP_016879361.1 | 2 - 3 | 133 | 61 | |
XM_017023873.1 | XP_016879362.1 | 2 - 3 | 300 | 61 | |
XM_017023874.1 | XP_016879363.1 | 2 - 3 | 133 | 61 | |
XM_017023876.1 | XP_016879365.1 | 2 - 3 | 178 | 61 | |
XM_017023877.1 | XP_016879366.1 | 2 - 3 | 133 | 61 | |
GenBank mRNA | AK022425.1 | - | 2 - 3 | 133 | 61 |
AL135642.1 | - | 2 - 3 | 133 | 61 | |
HY078299.1 | - | 2 - 3 | 145 | 61 |