The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000162.3 | NP_000153.1 | 3 - 4 | 830 | 103 |
NM_033507.1 | NP_277042.1 | 3 - 4 | 531 | 103 | |
NM_033508.1 | NP_277043.1 | 4 - 5 | 655 | 103 | |
XM_017011966.1 | XP_016867455.1 | 3 - 4 | 846 | 103 | |
GenBank mRNA | AK122876.1 | - | 3 - 4 | 525 | 103 |
BC001890.1 | - | 3 - 4 | 830 | 103 | |
BT007062.1 | - | 3 - 4 | 360 | 103 | |
EU008605.1 | - | 3 - 4 | 360 | 103 | |
M69051.1 | - | 4 - 5 | 642 | 103 | |
M88011.1 | - | 3 - 4 | 689 | 103 | |
M90299.1 | - | 3 - 4 | 818 | 103 |