Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NCAM1
Entrez Gene ID
4684
Gene Name
neural cell adhesion molecule 1
Gene Aliases
CD56, MSK39, NCAM
Chromosome Location
Chr.11: 112961247 - 113278436 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000615.6 NP_000606.3 19 - 19 5206 64
NM_001242607.1 NP_001229536.1 21 - 21 5314 64
NM_181351.4 NP_851996.2 20 - 20 5236 64
GenBank mRNA AK054570.1 - 1 - 1 653 64
AL832563.1 - 2 - 2 2852 64
BC014205.2 - 14 - 14 4100 64

View Full Product Details