The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145062.2 | NP_659499.2 | 5 - 6 | 1224 | 81 |
XM_011535565.2 | XP_011533867.1 | 5 - 6 | 1617 | 81 | |
XM_011535567.1 | XP_011533869.1 | 5 - 6 | 901 | 81 | |
XM_017010393.1 | XP_016865882.1 | 5 - 6 | 1196 | 81 | |
XM_017010394.1 | XP_016865883.1 | 4 - 5 | 650 | 81 | |
XM_017010395.1 | XP_016865884.1 | 6 - 7 | 1754 | 81 | |
XM_017010396.1 | XP_016865885.1 | 6 - 7 | 1134 | 81 | |
GenBank mRNA | AK054582.1 | - | 5 - 6 | 1210 | 81 |
AK307523.1 | - | 4 - 5 | 650 | 81 | |
BC016879.1 | - | 5 - 6 | 1192 | 81 | |
BX537872.1 | - | 5 - 6 | 1185 | 81 | |
KU178892.1 | - | 4 - 5 | 964 | 81 | |
KU178893.1 | - | 4 - 5 | 964 | 81 | |
KU178894.1 | - | 4 - 5 | 964 | 81 |