The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000722.3 | NP_000713.2 | 39 - 39 | 6875 | 148 |
XM_005250570.2 | XP_005250627.1 | 39 - 39 | 7018 | 148 | |
XM_005250572.2 | XP_005250629.1 | 39 - 39 | 6967 | 148 | |
XM_005250573.2 | XP_005250630.1 | 38 - 38 | 6958 | 148 | |
XM_005250574.2 | XP_005250631.1 | 38 - 38 | 6944 | 148 | |
XM_006716118.2 | XP_006716181.1 | 40 - 40 | 7041 | 148 | |
XM_006716119.3 | XP_006716182.1 | 40 - 40 | 8824 | 148 | |
XM_006716120.3 | XP_006716183.1 | 40 - 40 | 8266 | 148 | |
XM_006716121.3 | XP_006716184.1 | 22 - 22 | 5890 | 148 | |
XM_011516571.2 | XP_011514873.1 | 40 - 40 | 7027 | 148 | |
XM_011516572.2 | XP_011514874.1 | 39 - 39 | 7003 | 148 | |
XM_017012588.1 | XP_016868077.1 | 39 - 39 | 8208 | 148 | |
GenBank mRNA | AK055040.1 | - | 1 - 1 | 1341 | 148 |
AL833141.1 | - | 1 - 1 | 1137 | 148 |