The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308258.1 | NP_001295187.1 | 3 - 4 | 613 | 104 |
NM_032383.4 | NP_115759.2 | 4 - 5 | 1108 | 104 | |
XM_005247834.3 | XP_005247891.1 | 4 - 5 | 1083 | 104 | |
XM_017007323.1 | XP_016862812.1 | 4 - 5 | 1083 | 104 | |
GenBank mRNA | AK055245.1 | - | 4 - 5 | 1567 | 104 |
AK056575.1 | - | 3 - 4 | 376 | 104 | |
AY033141.1 | - | 4 - 5 | 1108 | 104 | |
BC022062.2 | - | 3 - 4 | 531 | 104 | |
BC040359.1 | - | 4 - 5 | 1045 | 104 |