The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267046.1 | NP_001253975.1 | 1 - 2 | 139 | 63 |
NM_152330.3 | NP_689543.1 | 1 - 2 | 139 | 63 | |
XM_017020967.1 | XP_016876456.1 | 1 - 2 | 89 | 63 | |
XM_017020968.1 | XP_016876457.1 | 1 - 2 | 89 | 63 | |
GenBank mRNA | AK055545.1 | - | 1 - 2 | 50 | 63 |
AK315764.1 | - | 1 - 2 | 60 | 63 | |
BC020521.2 | - | 1 - 2 | 87 | 63 | |
BX161430.1 | - | 1 - 2 | 139 | 63 | |
BX647863.1 | - | 1 - 2 | 51 | 63 |