The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030962.3 | NP_112224.1 | 38 - 39 | 5461 | 67 |
XM_005253154.4 | XP_005253211.1 | 39 - 40 | 5566 | 67 | |
XM_005253155.4 | XP_005253212.1 | 38 - 39 | 5434 | 67 | |
XM_011520394.2 | XP_011518696.1 | 38 - 39 | 5449 | 67 | |
XM_017018372.1 | XP_016873861.1 | 38 - 39 | 6855 | 67 | |
XM_017018373.1 | XP_016873862.1 | 39 - 40 | 5485 | 67 | |
XM_017018374.1 | XP_016873863.1 | 37 - 38 | 5341 | 67 | |
XM_017018375.1 | XP_016873864.1 | 37 - 38 | 5329 | 67 | |
GenBank mRNA | AK091362.1 | - | 6 - 7 | 797 | 67 |
AK123621.1 | - | 3 - 4 | 627 | 67 | |
AY234241.1 | - | 38 - 39 | 5427 | 67 | |
BC011143.1 | - | 1 - 2 | 77 | 67 | |
BC043389.1 | - | 4 - 5 | 505 | 67 | |
BC053867.1 | - | 4 - 5 | 539 | 67 | |
BC063656.1 | - | 4 - 5 | 468 | 67 | |
BC101466.1 | - | 38 - 39 | 5420 | 67 | |
BX538184.1 | - | 4 - 5 | 423 | 67 | |
BX647533.1 | - | 11 - 12 | 5038 | 67 | |
CR749312.1 | - | 12 - 13 | 1765 | 67 |