The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145244.3 | NP_660287.1 | 2 - 3 | 332 | 78 |
GenBank mRNA | AK094236.1 | - | 2 - 3 | 302 | 78 |
AK312926.1 | - | 2 - 3 | 302 | 78 | |
AL832277.1 | - | 2 - 3 | 284 | 78 | |
BC013592.2 | - | 2 - 3 | 290 | 78 | |
DB208869.1 | - | 2 - 3 | 332 | 78 |