The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005751.4 | NP_005742.4 | 24 - 25 | 6199 | 119 |
NM_147185.2 | NP_671714.1 | 24 - 25 | 6199 | 119 | |
XM_017011643.1 | XP_016867132.1 | 25 - 26 | 6303 | 119 | |
XM_017011646.1 | XP_016867135.1 | 25 - 26 | 6303 | 119 | |
XM_017011649.1 | XP_016867138.1 | 25 - 26 | 6303 | 119 | |
XM_017011650.1 | XP_016867139.1 | 24 - 25 | 6207 | 119 | |
XM_017011653.1 | XP_016867142.1 | 23 - 24 | 6114 | 119 | |
XM_017011654.1 | XP_016867143.1 | 23 - 24 | 6066 | 119 | |
XM_017011659.1 | XP_016867148.1 | 3 - 4 | 1174 | 119 | |
XM_017011660.1 | XP_016867149.1 | 3 - 4 | 1174 | 119 | |
GenBank mRNA | AF247727.4 | - | 23 - 24 | 5833 | 119 |
AJ131693.1 | - | 24 - 25 | 6199 | 119 | |
AL117418.1 | - | 3 - 4 | 1163 | 119 | |
AY528715.1 | - | 6 - 7 | 1046 | 119 |