The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001129896.2 | NP_001123368.1 | 6 - 7 | 851 | 58 |
NM_001174068.1 | NP_001167539.1 | 5 - 6 | 730 | 58 | |
NM_080737.2 | NP_542775.2 | 7 - 8 | 899 | 58 | |
XM_005262229.2 | XP_005262286.1 | 5 - 6 | 731 | 58 | |
XM_011531068.2 | XP_011529370.1 | 7 - 8 | 928 | 58 | |
XM_017029966.1 | XP_016885455.1 | 7 - 8 | 928 | 58 | |
XM_017029967.1 | XP_016885456.1 | 6 - 7 | 820 | 58 | |
XM_017029968.1 | XP_016885457.1 | 5 - 6 | 731 | 58 | |
XM_017029969.1 | XP_016885458.1 | 5 - 6 | 705 | 58 | |
XM_017029970.1 | XP_016885459.1 | 5 - 6 | 653 | 58 | |
XM_017029971.1 | XP_016885460.1 | 6 - 7 | 831 | 58 | |
GenBank mRNA | AK292588.1 | - | 6 - 7 | 820 | 58 |
AK313086.1 | - | 5 - 6 | 705 | 58 | |
AL832596.1 | - | 5 - 6 | 730 | 58 | |
BC014913.1 | - | 7 - 8 | 875 | 58 | |
BX537410.1 | - | 6 - 7 | 818 | 58 |