The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138790.3 | NP_620145.2 | 1 - 2 | 175 | 70 |
XM_017020965.1 | XP_016876454.1 | 1 - 2 | 112 | 70 | |
GenBank mRNA | AK295352.1 | - | 1 - 2 | 168 | 70 |
AY358843.1 | - | 1 - 2 | 141 | 70 | |
BC015003.1 | - | 1 - 2 | 104 | 70 | |
DC328186.1 | - | 1 - 2 | 168 | 70 |