The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100594.1 | NP_001094064.1 | 4 - 5 | 1212 | 98 |
NM_017719.4 | NP_060189.3 | 5 - 6 | 1274 | 98 | |
XM_005265245.3 | XP_005265302.1 | 6 - 7 | 1324 | 98 | |
XM_011533861.2 | XP_011532163.1 | 3 - 4 | 465 | 98 | |
GenBank mRNA | AK291360.1 | - | 6 - 7 | 1325 | 98 |
AK314376.1 | - | 4 - 5 | 1097 | 98 | |
BC071567.1 | - | 4 - 5 | 1186 | 98 | |
CR749621.1 | - | 5 - 6 | 1134 | 98 | |
D43636.2 | - | 3 - 4 | 946 | 98 |