The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017999.2 | NP_001017999.1 | 2 - 3 | 450 | 61 |
NM_001018000.3 | NP_001018000.1 | 2 - 3 | 639 | 61 | |
NM_001018001.2 | NP_001018001.1 | 2 - 3 | 440 | 61 | |
NM_015209.2 | NP_056024.1 | 2 - 3 | 702 | 61 | |
NM_201628.2 | NP_963922.2 | 2 - 3 | 702 | 61 | |
XM_005245795.4 | XP_005245852.1 | 4 - 5 | 957 | 61 | |
XM_011541074.2 | XP_011539376.1 | 4 - 5 | 964 | 61 | |
XM_011541075.2 | XP_011539377.1 | 2 - 3 | 620 | 61 | |
XM_011541076.2 | XP_011539378.1 | 2 - 3 | 765 | 61 | |
XM_011541077.2 | XP_011539379.1 | 2 - 3 | 1744 | 61 | |
XM_011541080.2 | XP_011539382.1 | 4 - 5 | 957 | 61 | |
XM_017000768.1 | XP_016856257.1 | 4 - 5 | 957 | 61 | |
XM_017000769.1 | XP_016856258.1 | 4 - 5 | 957 | 61 | |
XM_017000770.1 | XP_016856259.1 | 4 - 5 | 957 | 61 | |
XM_017000771.1 | XP_016856260.1 | 3 - 4 | 625 | 61 | |
XM_017000772.1 | XP_016856261.1 | 2 - 3 | 304 | 61 | |
GenBank mRNA | AB015329.1 | - | 1 - 2 | 97 | 61 |
AY505119.1 | - | 3 - 4 | 630 | 61 | |
AY505120.1 | - | 2 - 3 | 639 | 61 | |
AY505121.1 | - | 2 - 3 | 440 | 61 | |
AY505122.1 | - | 2 - 3 | 450 | 61 | |
BC035501.2 | - | 2 - 3 | 638 | 61 |