The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173500.3 | NP_775771.3 | 14 - 15 | 3709 | 86 |
XM_005254171.4 | XP_005254228.1 | 13 - 14 | 3843 | 86 | |
XM_005254173.4 | XP_005254230.1 | 13 - 14 | 3591 | 86 | |
XM_006720402.3 | XP_006720465.1 | 13 - 14 | 3318 | 86 | |
XM_006720403.3 | XP_006720466.1 | 14 - 15 | 3485 | 86 | |
XM_017021950.1 | XP_016877439.1 | 12 - 13 | 3334 | 86 | |
GenBank mRNA | AB020654.1 | - | 2 - 3 | 1474 | 86 |
AF525400.1 | - | 17 - 18 | 4888 | 86 | |
AK131372.1 | - | 14 - 15 | 3433 | 86 | |
BC071556.1 | - | 15 - 16 | 3483 | 86 |